accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50292-00-7

IUPAC Name: N-hydroxyoctanamide sodium
Synonyms: caprylhydroxamic acid sodium
Molecular Weight (g/mol): 182.22
Molecular Formula: C8H17NNaO2
InChi Key: UOYPLHWLPMJIJM-UHFFFAOYSA-N
SMILES: [Na].CCCCCCCC(=O)NO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)