accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50296-37-2

IUPAC Name: bromotris(dimethylamino)phosphanium; hexafluoro-λ⁵-phosphanuide
Synonyms: bromotris(dimethylamino)phosphanium hexafluorophosphate
Molecular Weight (g/mol): 388.07
Molecular Formula: C6H18BrF6N3P2
InChi Key: XELPBWPBGHCIKX-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CN(C)[P+](Br)(N(C)C)N(C)C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)