accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 503555-96-2

IUPAC Name: 5-{[(tert-butoxy)carbonyl]amino}-2-chlorobenzoic acid
Synonyms: 5-[(tert-butoxycarbonyl)amino]-2-chlorobenzoic acid
Molecular Weight (g/mol): 271.7
Molecular Formula: C12H14ClNO4
InChi Key: QNRXWUBPCIBQMX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC(C(O)=O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results