accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClClCl

CAS RN 50375-10-5

IUPAC Name: 1,2,4-trichloro-3-methoxybenzene
Synonyms: 2,3,6-trichloroanisole
Molecular Weight (g/mol): 211.47
Molecular Formula: C7H5Cl3O
InChi Key: OTFNCXLUCRUNCH-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results