accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHSOONOO

CAS RN 50393-59-4

IUPAC Name: N-(naphthalen-1-yl)-2-nitrobenzene-1-sulfonamide
Synonyms: N-(naphthalen-1-yl)-2-nitrobenzenesulfonamide
Molecular Weight (g/mol): 328.34
Molecular Formula: C16H12N2O4S
InChi Key: YPAFWESEDDOFFH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1S(=O)(=O)NC1=CC=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results