accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHN

CAS RN 504-30-3

IUPAC Name: 2,3-dihydropyridazin-3-one
Synonyms: 2H-pyridazin-3-one
Molecular Weight (g/mol): 96.09
Molecular Formula: C4H4N2O
InChi Key: AAILEWXSEQLMNI-UHFFFAOYSA-N
SMILES: O=C1NN=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results