accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3CH3CH3

CAS RN 504-44-9

IUPAC Name: 2,6,11,15-tetramethylhexadecane
Synonyms: 2,6,11,15-tetramethylhexadecane
Molecular Weight (g/mol): 282.56
Molecular Formula: C20H42
InChi Key: KKFZXXATNGJPJS-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)CCCCC(C)CCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results