accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 504-53-0

IUPAC Name: pentatriacontan-18-one
Synonyms: 18-pentatriacontanone
Molecular Weight (g/mol): 506.94
Molecular Formula: C35H70O
InChi Key: DMCJFWXGXUEHFD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)CCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)