accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 504-88-1

IUPAC Name: 3-nitropropanoic acid
Synonyms: β-nitropropionic acid bovinocidin
Molecular Weight (g/mol): 119.08
Molecular Formula: C3H5NO4
InChi Key: WBLZUCOIBUDNBV-UHFFFAOYSA-N
SMILES: OC(=O)CC[N+]([O-])=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results