accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CCH3NH3

CAS RN 5041-09-8

IUPAC Name: 2-methylpropan-1-aminium chloride
Synonyms: 2-methylpropan-1-aminium chloride
Molecular Weight (g/mol): 109.6
Molecular Formula: C4H12ClN
InChi Key: BSMNBEHEFWDHJD-UHFFFAOYSA-N
SMILES: [Cl-].CC(C)C[NH3+]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results