CAS RN 5044-23-5
IUPAC Name:
1-(4-chlorophenyl)-2,5-dimethyl-1H-pyrrole
Synonyms:
1-(4-chlorophenyl)-2,5-dimethylpyrrole
Molecular Weight (g/mol):
205.69
Molecular Formula:
C12H12ClN
InChi Key:
NNKFIFOSLJJRES-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C)N1C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
1-(4-Chlorophenyl)-2,5-dimethylpyrrole 98.0+%, TCI America™
CAS: 5044-23-5 Molecular Formula: C12H12ClN Molecular Weight (g/mol): 205.685 InChI Key: NNKFIFOSLJJRES-UHFFFAOYSA-N Synonym: 2,5-Dimethyl-1-(4-chlorophenyl)pyrrole PubChem CID: 78742 IUPAC Name: 1-(4-chlorophenyl)-2,5-dimethylpyrrole SMILES: CC1=CC=C(N1C2=CC=C(C=C2)Cl)C