accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CF(E/Z)

CAS RN 504433-86-7

IUPAC Name: 2-[2-(4-fluorophenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[2-(4-fluorophenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 248.1
Molecular Formula: C14H18BFO2
InChi Key: ZJRAXVMUDOVAOD-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C=CC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results