accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50463-48-4

IUPAC Name: 3-[4-(benzyloxy)phenyl]propanoic acid
Synonyms: 3-[4-(benzyloxy)phenyl]propanoic acid
Molecular Weight (g/mol): 256.3
Molecular Formula: C16H16O3
InChi Key: QTSAUVQZNADEKS-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results