accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOClClH2C

CAS RN 50471-44-8

IUPAC Name: 3-(3,5-dichlorophenyl)-5-ethenyl-5-methyl-1,3-oxazolidine-2,4-dione
Synonyms: vinclozolin epa pesticide chemical code 113201 C12H9cl2NO3 vinclozoline vorlan caswell no. 323C ornalin vinchlozoline ronilan
Molecular Weight (g/mol): 286.11
Molecular Formula: C12H9Cl2NO3
InChi Key: FSCWZHGZWWDELK-UHFFFAOYNA-N
SMILES: CC1(OC(=O)N(C1=O)C1=CC(Cl)=CC(Cl)=C1)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results