accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2

CAS RN 5048-26-0

IUPAC Name: hex-5-en-1-yl acetate
Synonyms: hex-5-en-1-yl acetate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: MPLWNENKBSBMFN-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCC=C
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)