accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2

CAS RN 5048-35-1

IUPAC Name: oct-7-en-1-yl acetate
Synonyms: oct-7-en-1-yl acetate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: DBRVFNFZYZFKRC-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results