accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHHCl

CAS RN 50492-22-3

IUPAC Name: azepan-4-one hydrochloride
Synonyms: azepan-4-one hydrochloride
Molecular Weight (g/mol): 149.62
Molecular Formula: C6H12ClNO
InChi Key: WFTRLIZPJMFJER-UHFFFAOYSA-N
SMILES: Cl.O=C1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results