accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3H3COHCH2

CAS RN 505-32-8

IUPAC Name: 3,7,11,15-tetramethylhexadec-1-en-3-ol
Synonyms: isophytol tetramethyl-1-hexadecen-3-ol
Molecular Weight (g/mol): 296.54
Molecular Formula: C20H40O
InChi Key: KEVYVLWNCKMXJX-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)CCCC(C)CCCC(C)(O)C=C
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)