accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO

CAS RN 505-48-6

IUPAC Name: octanedioic acid
Synonyms: suberic acid octenedioic acid oktandisaeure monosodium salt suberic acid suberinsaeure cis-octenedioic acid dipotassium salt suberic acid suberic acid (substance) disodium salt suberic acid cork acid
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: TYFQFVWCELRYAO-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCC(O)=O
Percent Purity 
  • (1)
  • (3)
  • (3)
  • (11)
Quantity 
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
Boiling Point 
  • (7)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (9)
Physical Form 
  • (3)
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (4)
  • (5)
  • (3)
  • (6)
  • (5)
  • (1)
  • (3)
  • (3)
  • (11)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (7)
  • (3)
  • (3)
  • (4)
  • (9)
  • (3)
  • (5)
  • (3)