accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 505084-55-9

NClFFFI

CAS RN 505084-55-9

IUPAC Name: 2-chloro-4-iodo-5-(trifluoromethyl)pyridine
Synonyms: 2-chloro-4-iodo-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 307.44
Molecular Formula: C6H2ClF3IN
InChi Key: XEEQBBXEZSNIQV-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(Cl)C=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results