accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50528-86-4

IUPAC Name: 1-chloro-2-isocyanato-4-(trifluoromethyl)benzene
Synonyms: 1-chloro-2-isocyanato-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 221.56
Molecular Formula: C8H3ClF3NO
InChi Key: WEPYOPYMWSHRIW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)C(=C1)N=C=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)