accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBr

CAS RN 50530-12-6

IUPAC Name: 10-bromodecanoic acid
Synonyms: 10-bromodecanoic acid
Molecular Weight (g/mol): 251.16
Molecular Formula: C10H19BrO2
InChi Key: PGVRSPIEZYGOAD-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCBr
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)