CAS RN 50551-58-1
IUPAC Name:
ethyl 7-methoxy-1-benzofuran-2-carboxylate
Synonyms:
ethyl 7-methoxy-1-benzofuran-2-carboxylate
Molecular Weight (g/mol):
220.22
Molecular Formula:
C12H12O4
InChi Key:
SXCMZYVXJUJIDJ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC2=CC=CC(OC)=C2O1
1
–
1
of
1
results
1
Promotions Available
Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%
Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%
CAS: 50551-58-1 Molecular Formula: C12H12O4 Molecular Weight (g/mol): 220.22 MDL Number: MFCD00060510 InChI Key: SXCMZYVXJUJIDJ-UHFFFAOYSA-N PubChem CID: 521284 IUPAC Name: ethyl 7-methoxy-1-benzofuran-2-carboxylate SMILES: CCOC(=O)C1=CC2=CC=CC(OC)=C2O1