accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHO

CAS RN 5057-98-7

IUPAC Name: cyclopentane-1,2-diol
Synonyms: cyclopentane-1,2-diol
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: VCVOSERVUCJNPR-UHFFFAOYNA-N
SMILES: OC1CCCC1O
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)