accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOClFFFNOO

CAS RN 50594-66-6

IUPAC Name: 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoic acid
Synonyms: acifluorfen
Molecular Weight (g/mol): 361.66
Molecular Formula: C14H7ClF3NO5
InChi Key: NUFNQYOELLVIPL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(OC2=CC=C(C=C2Cl)C(F)(F)F)=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results