accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50594-67-7

IUPAC Name: methyl 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoate
Synonyms: acifluorfen-ME
Molecular Weight (g/mol): 375.68
Molecular Formula: C15H9ClF3NO5
InChi Key: AHGMXAFUHVRQAD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC2=CC=C(C=C2Cl)C(F)(F)F)=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results