accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO(Z)

CAS RN 506-37-6

IUPAC Name: (15Z)-tetracos-15-enoic acid
Synonyms: nervonic acid nervonic acid (substance) nervonoyl acid selacholeic acid selacholeate cis-15-teracosenoic acid cis-selacholeate nervonsaeure cis-selacholeic acid cis-15-teracosenoate
Molecular Weight (g/mol): 366.63
Molecular Formula: C24H46O2
InChi Key: GWHCXVQVJPWHRF-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCCCCCCC(O)=O
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)