CAS RN 506-38-7
IUPAC Name:
pentacosanoic acid
Synonyms:
pentacosanoic acid hyenate hyenic acid
Molecular Weight (g/mol):
382.67
Molecular Formula:
C25H50O2
InChi Key:
MWMPEAHGUXCSMY-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
1
–
1
of
1
results
1
Pentacosanoic Acid 96.0+%, TCI America™
CAS: 506-38-7 Molecular Formula: C25H50O2 Molecular Weight (g/mol): 382.67 MDL Number: MFCD00020551 InChI Key: MWMPEAHGUXCSMY-UHFFFAOYSA-N PubChem CID: 10468 ChEBI: CHEBI:39420 IUPAC Name: pentacosanoic acid SMILES: CCCCCCCCCCCCCCCCCCCCCCCCC(O)=O