accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH(E)

CAS RN 506-42-3

IUPAC Name: (9E)-octadec-9-en-1-ol
Synonyms: (9E)-octadec-9-en-1-ol
Molecular Weight (g/mol): 268.49
Molecular Formula: C18H36O
InChi Key: ALSTYHKOOCGGFT-MDZDMXLPSA-N
SMILES: CCCCCCCC\C=C\CCCCCCCCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results