accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 506-46-7

IUPAC Name: hexacosanoic acid
Synonyms: hexacosanoic acid c26:0 aliphatic carboxylic acid hexacosansaeure aliphatic carboxylic acid, c26:0 hexaeicosanoic acid ceratinic acid cerotic acid (substance) cerinic acid cerylic acid hexacosanoate (n-C26:0)
Molecular Weight (g/mol): 396.7
Molecular Formula: C26H52O2
InChi Key: XMHIUKTWLZUKEX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)