accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 506-50-3

IUPAC Name: triacontanoic acid
Synonyms: triacontanoic acid myricic acid mellisic acid melissic acid triacontansaeure
Molecular Weight (g/mol): 452.81
Molecular Formula: C30H60O2
InChi Key: VHOCUJPBKOZGJD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)