accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 506-78-5

IUPAC Name: carbononitridic iodide
Synonyms: cyanogen iodide
Molecular Weight (g/mol): 152.92
Molecular Formula: CIN
InChi Key: WPBXOELOQKLBDF-UHFFFAOYSA-N
SMILES: IC#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)