accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CClO

CAS RN 50606-95-6

IUPAC Name: 4-hexylbenzoyl chloride
Synonyms: 4-hexylbenzoyl chloride
Molecular Weight (g/mol): 224.73
Molecular Formula: C13H17ClO
InChi Key: XRAHLPNMIIAEPP-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)