accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOO

CAS RN 5061-21-2

IUPAC Name: 3-bromooxolan-2-one
Synonyms: 3-bromooxolan-2-one
Molecular Weight (g/mol): 164.99
Molecular Formula: C4H5BrO2
InChi Key: LFJJGHGXHXXDFT-UHFFFAOYNA-N
SMILES: BrC1CCOC1=O
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)