accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CO(S)OH(S)HOO

CAS RN 50622-09-8

IUPAC Name: [(4S,5S)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
Synonyms: [(4S,5S)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
Molecular Weight (g/mol): 162.19
Molecular Formula: C7H14O4
InChi Key: INVRLGIKFANLFP-WDSKDSINSA-N
SMILES: CC1(C)O[C@@H](CO)[C@H](CO)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results