accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50638-50-1

IUPAC Name: 4-bromo-N,N,3-trimethylaniline
Synonyms: 4-bromo-N,N,3-trimethylaniline
Molecular Weight (g/mol): 214.11
Molecular Formula: C9H12BrN
InChi Key: DPFVFQHTPIFECX-UHFFFAOYSA-N
SMILES: CN(C)C1=CC(C)=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results