accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50638-54-5

IUPAC Name: 4-bromo-N,N,2,6-tetramethylaniline
Synonyms: 4-bromo-N,N,2,6-tetramethylaniline
Molecular Weight (g/mol): 228.13
Molecular Formula: C10H14BrN
InChi Key: NDDKEDQKCHIHMI-UHFFFAOYSA-N
SMILES: CN(C)C1=C(C)C=C(Br)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)