accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOO

CAS RN 50662-95-8

IUPAC Name: 4-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 4-[(1,3-dioxo-2-benzofuran-5-yl)oxy]-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 310.22
Molecular Formula: C16H6O7
InChi Key: OPVHOFITDJSMOD-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C1C=CC(OC1=CC=CC3=C1C(=O)OC3=O)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results