accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50667-69-1

IUPAC Name: 2,2,2-trifluoro-N-(hydroxymethyl)acetamide
Synonyms: 2,2,2-trifluoro-N-(hydroxymethyl)acetamide
Molecular Weight (g/mol): 143.07
Molecular Formula: C3H4F3NO2
InChi Key: URUWXKFAEKTWKG-UHFFFAOYSA-N
SMILES: OCNC(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results