accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50670-50-3

IUPAC Name: 4'-methyl-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-methyl-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: QIBWMVSMTSYUSK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results