accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNH2HOO

CAS RN 50670-83-2

IUPAC Name: 2-amino-5-acetamidobenzoic acid
Synonyms: 2-amino-5-acetamidobenzoic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C9H10N2O3
InChi Key: GSOHXJQXAKNJES-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(N)C(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results