accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50687-62-2

CAS RN 50687-62-2

IUPAC Name: 4-methoxyphenyl 4-hydroxybenzoate
Synonyms: 4-methoxyphenyl 4-hydroxybenzoate
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: MTQSVJUCLQYISS-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC(=O)C2=CC=C(O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)