accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 507-36-8

IUPAC Name: 2-bromo-2-methylbutane
Synonyms: butane, 2-bromo-2-methyl-
Molecular Weight (g/mol): 151.05
Molecular Formula: C5H11Br
InChi Key: JOUWCKCVTDSMHF-UHFFFAOYSA-N
SMILES: CCC(C)(C)Br
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)