CAS RN 50703-06-5
IUPAC Name:
hydrogen methyl 1,3-thiazolidine-4-carboxylate chloride
Synonyms:
hydrogen methyl 1,3-thiazolidine-4-carboxylate chloride
Molecular Weight (g/mol):
183.65
Molecular Formula:
C5H10ClNO2S
InChi Key:
YEFOSJYXVLNYSL-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].COC(=O)C1CSCN1
1
–
1
of
1
results
1
Promotions Available
Methyl thiazolidine-2-carboxylate hydrochloride, 98%
Methyl thiazolidine-2-carboxylate hydrochloride, 98%
CAS: 50703-06-5 Molecular Formula: C5H10ClNO2S Molecular Weight (g/mol): 183.65 MDL Number: MFCD00068022 InChI Key: YEFOSJYXVLNYSL-UHFFFAOYNA-N Synonym: 2-thiazolidinecarboxylic acid methyl ester,2-thiazolidinecarboxylic acid, methyl ester,thiazolidine-2-carboxylic acid methyl ester,methyl thiazolidine-2-carboxylate,+-thiazolidine-2-carboxylic acid methyl ester,+/--thiazolidine-2-carboxylic acid methyl ester,2-thiazolidinecarboxylic acid, methyl ester 6ci,8ci,9ci PubChem CID: 2737507 IUPAC Name: methyl 1,3-thiazolidine-2-carboxylate SMILES: [H+].[Cl-].COC(=O)C1CSCN1