CAS RN 5071-61-4
IUPAC Name:
3-phenyl-1H-pyrazole-5-carboxylate
Synonyms:
5-phenyl-2H-pyrazole-3-carboxylate
Molecular Weight (g/mol):
187.18
Molecular Formula:
C10H7N2O2
InChi Key:
QBPUOAJBMXXBNU-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC(=NN1)C1=CC=CC=C1
1
–
1
of
1
results
1
3-Phenylpyrazole-5-carboxylic Acid Hydrate 97.0+%, TCI America™
CAS: 5071-61-4 Molecular Formula: C10H7N2O2 Molecular Weight (g/mol): 187.18 MDL Number: MFCD01248821,MFCD05170017 InChI Key: QBPUOAJBMXXBNU-UHFFFAOYSA-M PubChem CID: 121025 IUPAC Name: 3-phenyl-1H-pyrazole-5-carboxylate SMILES: [O-]C(=O)C1=CC(=NN1)C1=CC=CC=C1