accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SHNONH2OHO

CAS RN 5072-26-4

IUPAC Name: 2-amino-4-[butyl(imino)oxo-λ⁶-sulfanyl]butanoic acid
Synonyms: buthionine sulfoximine|buthionine-S,R-sulfoximine|buthionine sulphoximine|buthionine sulfoxamine|buthione sulfoximine|sulfoximine, buthionine|butionine sulfoximine
Molecular Weight (g/mol): 222.3
Molecular Formula: C8H18N2O3S
InChi Key: KJQFBVYMGADDTQ-UHFFFAOYNA-N
SMILES: CCCCS(=N)(=O)CCC(N)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)