accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2NNNNBrBr(E/Z)

CAS RN 50768-79-1

IUPAC Name: 4-[2-(3,5-dibromopyridin-2-yl)diazen-1-yl]benzene-1,3-diamine
Synonyms: 4-[2-(3,5-dibromopyridin-2-yl)diazen-1-yl]benzene-1,3-diamine
Molecular Weight (g/mol): 371.04
Molecular Formula: C11H9Br2N5
InChi Key: NDSPKICAQDKJTG-UHFFFAOYSA-N
SMILES: NC1=CC(N)=C(C=C1)N=NC1=C(Br)C=C(Br)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results