accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50783-80-7

IUPAC Name: 3-(dimethylamino)-6-[2-(pyridin-2-yl)hydrazin-1-ylidene]cyclohexa-2,4-dien-1-one
Synonyms: 3-(dimethylamino)-6-[2-(pyridin-2-yl)hydrazin-1-ylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 242.28
Molecular Formula: C13H14N4O
InChi Key: HHPYHNOAMVRXBT-UHFFFAOYSA-N
SMILES: CN(C)C1=CC(=O)C(C=C1)=NNC1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results