accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50793-85-6

IUPAC Name: 4-cyanophenyl 4-hexylbenzoate
Synonyms: 4-cyanophenyl 4-hexylbenzoate
Molecular Weight (g/mol): 307.39
Molecular Formula: C20H21NO2
InChi Key: DEUWEGPRKHPNKB-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C(=O)OC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results