accessibility menu, dialog, popup

UserName

Viewing profile as user:
POOO∗∗Nan

CAS RN 50813-16-6

IUPAC Name: Sodium polyphosphate
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (NaO3P)n
InChi Key: —
SMILES: [Na]OP(-*)(=O)O-*
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)